Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium Sulfate, MP Biomedicals

Catalog No. ICN227135 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN227135 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN227135 Supplier MP Biomedicals, Inc Supplier No. 0522713580

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34806-73-0
Molecular Formula H2O9S2Zr
Molecular Weight (g/mol) 301.35
MDL Number MFCD00167043
InChI Key UXRMCZYAVOHQNB-UHFFFAOYSA-J
Synonym zirconium iv sulfate hydrate, zr so4 2 hydrate, 2so4.zr.h2o, zirconium 4+ hydrate disulfate, zirconium 4+ ion hydrate disulfate, sulfuric acid,zirconium 4+ salt 2:1 , hydrate 9ci, zirconium iv sulfate hydrate trace metals basis
PubChem CID 53249516
IUPAC Name zirconium(4+) hydrate disulfate
SMILES O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Quantity 100 g
Formula Weight 355.41
Chemical Name or Material Zirconium Sulfate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.