accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 3481-20-7

IUPAC Name: 2,3,5,6-tetrachloroaniline
Synonyms: 2,3,5,6-tetrachloroaniline
Molecular Weight (g/mol): 230.9
Molecular Formula: C6H3Cl4N
InChi Key: YTDHEFNWWHSXSU-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C(Cl)=CC(Cl)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results