Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrachloroaniline 98.0+%, TCI America™
SDP

Catalog No. T11271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T11271G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T11271G Supplier TCI America Supplier No. T11271G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3481-20-7
Molecular Formula C6H3Cl4N
Molecular Weight (g/mol) 230.897
MDL Number MFCD00007659
InChI Key YTDHEFNWWHSXSU-UHFFFAOYSA-N
Synonym benzenamine, 2,3,5,6-tetrachloro, aniline, 2,3,5,6-tetrachloro, unii-48a7xff90t, 2,3,5,6-tetrachlorophenylamine, 2,5,6-tetrachloroaniline, aniline,3,5,6-tetrachloro, benzenamine,3,5,6-tetrachloro, ytdhefnwwhsxsu-uhfffaoysa, aniline, 2,3,5,6-tetrachloro-8ci, benzenamine, 2,3,5,6-tetrachloro-9ci
PubChem CID 18998
IUPAC Name 2,3,5,6-tetrachloroaniline
SMILES C1=C(C(=C(C(=C1Cl)Cl)N)Cl)Cl

Specifications

Melting Point 109°C
Quantity 1 g
Physical Form Crystal-Powder at 20°C
UN Number 3077
Formula Weight 230.90
Percent Purity ≥98.0% (GC)
Chemical Name or Material 2,3,5,6-Tetrachloroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.