accessibility menu, dialog, popup

UserName

Viewing profile as user:
HO(R)O(S)O(S)(R)HO(R)OH(S)O(R)HOONOO(R)OH(S)HO(S)HO

CAS RN 3482-57-3

IUPAC Name: (2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: (2S,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6S)-4,5-dihydroxy-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Weight (g/mol): 463.39
Molecular Formula: C18H25NO13
InChi Key: IAYJZWFYUSNIPN-KFRZSCGFSA-N
SMILES: OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](OC3=CC=C(C=C3)[N+]([O-])=O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@@H]1O
Grade 
  • (1)
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)