accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34822-90-7

IUPAC Name: (cyclopenta-2,4-dien-1-yl)thallane
Synonyms: cyclopentadienylthallium
Molecular Weight (g/mol): 269.48
Molecular Formula: C5H5Tl
InChi Key: RKLJDPOVAWGBSD-UHFFFAOYSA-N
SMILES: [Tl]C1C=CC=C1
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)