Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cyclopentadienyl Thallium 95.0+%, TCI America™
SDP

Catalog No. C09545G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09545G 5 g
C09541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09545G Supplier TCI America Supplier No. C09545G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 34822-90-7
Molecular Formula C5H5Tl
MDL Number MFCD00134141

Specifications

Physical Form Crystalline Powder
Quantity 5 g
UN Number 1707
Formula Weight 269.48
Percent Purity ≥95.0% (T)
Chemical Name or Material Cyclopentadienyl Thallium
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.