accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34824-20-9

IUPAC Name: [8-(hydroxymethyl)anthracen-1-yl]methanol
Synonyms: [8-(hydroxymethyl)anthracen-1-yl]methanol
Molecular Weight (g/mol): 238.29
Molecular Formula: C16H14O2
InChi Key: GSHJWFSWKUADLL-UHFFFAOYSA-N
SMILES: OCC1=CC=CC2=CC3=CC=CC(CO)=C3C=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results