Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,8-Bis(hydroxymethyl)anthracene 98.0+%, TCI America™
SDP

Catalog No. B19041G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19041G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B19041G Supplier TCI America Supplier No. B19041G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34824-20-9
Molecular Formula C16H14O2
Molecular Weight (g/mol) 238.286
MDL Number MFCD01321137
InChI Key GSHJWFSWKUADLL-UHFFFAOYSA-N
PubChem CID 12969385
IUPAC Name [8-(hydroxymethyl)anthracen-1-yl]methanol
SMILES C1=CC2=CC3=C(C=C2C(=C1)CO)C(=CC=C3)CO

Specifications

Melting Point 222°C
Quantity 1 g
Formula Weight 238.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,8-Bis(hydroxymethyl)anthracene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.