accessibility menu, dialog, popup

UserName

Viewing profile as user:
PN

CAS RN 34825-99-5

IUPAC Name: 2-(diphenylphosphanyl)benzonitrile
Synonyms: 2-(diphenylphosphanyl)benzonitrile
Molecular Weight (g/mol): 287.3
Molecular Formula: C19H14NP
InChi Key: FEOUZPAYKDNQBW-UHFFFAOYSA-N
SMILES: N#CC1=CC=CC=C1P(C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results