Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(Diphenylphosphino)benzonitrile 98.0+%, TCI America™
SDP

Catalog No. D4953200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4953200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4953200MG Supplier TCI America Supplier No. D4953200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34825-99-5
Molecular Formula C19H14NP
Molecular Weight (g/mol) 287.302
MDL Number MFCD05864011
InChI Key FEOUZPAYKDNQBW-UHFFFAOYSA-N
Synonym (2-Cyanophenyl)diphenylphosphine
PubChem CID 4590862
IUPAC Name 2-diphenylphosphanylbenzonitrile
SMILES C1=CC=C(C=C1)P(C2=CC=CC=C2)C3=CC=CC=C3C#N

Specifications

Melting Point 150°C
Color Yellow
Quantity 200 mg
Formula Weight 287.30
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(Diphenylphosphino)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.