accessibility menu, dialog, popup

UserName

Viewing profile as user:
(S)OHNOHOCH3O

CAS RN 3483-82-7

IUPAC Name: ethyl (2S)-3-(4-hydroxyphenyl)-2-(phenylformamido)propanoate
Synonyms: ethyl N-benzoyl-L-tyrosinate
Molecular Weight (g/mol): 313.35
Molecular Formula: C18H19NO4
InChi Key: SRLROPAFMUDDRC-XISACWJONA-N
SMILES: CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)