Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Benzoyl-L-tyrosine Ethyl Ester 98.0+%, TCI America™
SDP

Catalog No. B11405G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11405G 5 g
B11401G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11405G Supplier TCI America Supplier No. B11405G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 3483-82-7
Molecular Formula C18H19NO4
Molecular Weight (g/mol) 313.35
MDL Number MFCD00002388
InChI Key SRLROPAFMUDDRC-XISACWJONA-N
Synonym n-benzoyl-l-tyrosine ethyl ester, bz-tyr-oet, ethyl n-benzoyl-l-tyrosinate, btee, ethyl benzoyltyrosinate, benzoyltyrosine ethyl ester, benzoyl-l-tyrosine ethyl ester, s-ethyl 2-benzamido-3-4-hydroxyphenyl propanoate, benzoyl-tyr-oet, l-tyrosine, n-benzoyl-, ethyl ester
PubChem CID 77033
IUPAC Name ethyl (2S)-3-(4-hydroxyphenyl)-2-(phenylformamido)propanoate
SMILES CCOC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)C1=CC=CC=C1

Specifications

Melting Point 122°C
Color White
Quantity 5 g
Formula Weight 313.35
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Benzoyl-L-tyrosine Ethyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.