accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClCl

CAS RN 34883-41-5

IUPAC Name: 3,5-dichloro-1,1'-biphenyl
Synonyms: 3,5-dichlorobiphenyl
Molecular Weight (g/mol): 223.1
Molecular Formula: C12H8Cl2
InChi Key: QHZSDTDMQZPUKC-UHFFFAOYSA-N
SMILES: ClC1=CC(=CC(Cl)=C1)C1=CC=CC=C1
Solvent or Matrix 
  • (1)
  • (1)
Element 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)