accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NN

CAS RN 34884-10-1

IUPAC Name: 1-methyl-1H-pyrrole-2-carbonitrile
Synonyms: 1-methylpyrrole-2-carbonitrile
Molecular Weight (g/mol): 106.13
Molecular Formula: C6H6N2
InChi Key: JRQSGIQEBOZPHK-UHFFFAOYSA-N
SMILES: CN1C=CC=C1C#N
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results