Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Methylpyrrole-2-carbonitrile 98.0+%, TCI America™
SDP

Catalog No. M27845G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27845G 5 g
M27841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27845G Supplier TCI America Supplier No. M27845G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34884-10-1
Molecular Formula C6H6N2
Molecular Weight (g/mol) 106.128
MDL Number MFCD02693721
InChI Key JRQSGIQEBOZPHK-UHFFFAOYSA-N
Synonym 2-Cyano-1-methylpyrrole
PubChem CID 2064111
IUPAC Name 1-methylpyrrole-2-carbonitrile
SMILES CN1C=CC=C1C#N

Specifications

Color Yellow
Boiling Point 87°C
Quantity 5 g
Formula Weight 106.13
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Methylpyrrole-2-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.