accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 34896-80-5

IUPAC Name: 1-bromo-3-methanesulfonylbenzene
Synonyms: 1-bromo-3-methanesulfonylbenzene
Molecular Weight (g/mol): 235.1
Molecular Formula: C7H7BrO2S
InChi Key: WBOMXUMQOVQNKT-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=CC(Br)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results