accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 349091-53-8

IUPAC Name: N-benzyl-N-tert-butyl-3-nitrobenzamide
Synonyms: N-benzyl-N-tert-butyl-3-nitrobenzamide
Molecular Weight (g/mol): 312.37
Molecular Formula: C18H20N2O3
InChi Key: NOSAZZBVBDAHIZ-UHFFFAOYSA-N
SMILES: CC(C)(C)N(CC1=CC=CC=C1)C(=O)C1=CC(=CC=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results