CAS RN 34937-00-3
IUPAC Name:
hydrogen 11-(2-ethenylphenyl)-3,3-dimethoxy-2-oxa-7,10-diaza-3-silaundecane chloride
Synonyms:
hydrogen 11-(2-ethenylphenyl)-3,3-dimethoxy-2-oxa-7,10-diaza-3-silaundecane chloride
Molecular Weight (g/mol):
374.98
Molecular Formula:
C17H31ClN2O3Si
InChi Key:
MYWJOHLSXLTXCV-UHFFFAOYSA-N
SMILES:
[H+].[Cl-].CO[Si](CCCNCCNCC1=CC=CC=C1C=C)(OC)OC
1
–
1
of
1
results
1
N-[2-(N-Vinylbenzylamino)ethyl]-3-aminopropyltrimethoxysilane Hydrochloride (30-40% in Methanol), TCI America™
CAS: 34937-00-3 Molecular Formula: C17H30N2O3Si MDL Number: MFCD00060149 Synonym: Trimethoxy-N-[2-(N-vinylbenzylamino)ethyl]aminopropylsilane Hydrochloride