Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-[2-(N-Vinylbenzylamino)ethyl]-3-aminopropyltrimethoxysilane Hydrochloride (30-40% in Methanol), TCI America™
SDP

Catalog No. V005025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
V005025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. V005025ML Supplier TCI America Supplier No. V005025ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 34937-00-3
Molecular Formula C17H30N2O3Si
MDL Number MFCD00060149
Synonym Trimethoxy-N-[2-(N-vinylbenzylamino)ethyl]aminopropylsilane Hydrochloride

Specifications

Quantity 25 mL
UN Number 1230
Formula Weight 374.98
Physical Form Liquid
Chemical Name or Material N-[2-(N-Vinylbenzylamino)ethyl]-3-aminopropyltrimethoxysilane Hydrochloride (30-40% in Methanol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.