CAS RN 349438-38-6
IUPAC Name:
N-[(2H-1,3-benzodioxol-5-yl)methyl]-2,6-dichlorobenzamide
Synonyms:
N-(2H-1,3-benzodioxol-5-ylmethyl)-2,6-dichlorobenzamide
Molecular Weight (g/mol):
324.16
Molecular Formula:
C15H11Cl2NO3
InChi Key:
NMKJFZCBCIUYHI-UHFFFAOYSA-N
SMILES:
ClC1=CC=CC(Cl)=C1C(=O)NCC1=CC2=C(OCO2)C=C1
1
–
2
of
2
results
1
MilliporeSigma™ Calbiochem™ ALDH2 Agonist, Alda-1
The ALDH2 Agonist, Alda-1, also referenced under CAS 349438-38-6, controls the biological activity of ALDH2. This small molecule/inhibitor is primarily used for Activators/Inducers applications.
2
R&D Systems™ Alda 1
CAS: 349438-38-6 Molecular Formula: C15H11Cl2NO3