accessibility menu, dialog, popup

UserName

Viewing profile as user:
CuSHOOFFFSHOOFFF

CAS RN 34946-82-2

IUPAC Name: copper(II) triflate
Synonyms: copper(II) triflate
Molecular Weight (g/mol): 331.69
Molecular Formula: C2H2CuF6O4S2
InChi Key: TXWAGEFXPDWAQH-UHFFFAOYSA-N
SMILES: [Cu].FC(F)(F)S(=O)=O.FC(F)(F)S(=O)=O
Molecular Weight (g/mol) 
  • (8)
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (5)
Physical Form 
  • (4)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (2)
  • (8)
  • (8)
  • (2)
  • (2)
  • (1)
  • (2)
  • (4)
  • (4)
  • (1)
  • (2)
  • (3)
  • (5)
  • (4)
  • (3)
  • (2)