accessibility menu, dialog, popup

UserName

Viewing profile as user:
FCH2

CAS RN 350-51-6

IUPAC Name: 1-ethenyl-3-fluorobenzene
Synonyms: 1-ethenyl-3-fluorobenzene
Molecular Weight (g/mol): 122.14
Molecular Formula: C8H7F
InChi Key: ZJSKEGAHBAHFON-UHFFFAOYSA-N
SMILES: FC1=CC=CC(C=C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results