Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Fluorostyrene (stabilized with TBC) 97.0+%, TCI America™
SDP

Catalog No. F04095G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F04095G 5 g
F04091G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F04095G Supplier TCI America Supplier No. F04095G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 350-51-6
Molecular Formula C8H7F
Molecular Weight (g/mol) 122.142
MDL Number MFCD00000341
InChI Key ZJSKEGAHBAHFON-UHFFFAOYSA-N
Synonym 3-fluorostyrene, 1-fluoro-3-vinylbenzene, m-fluorostyrene, benzene, 1-ethenyl-3-fluoro, 1-ethenyl-3-fluoro-benzene, 3-fluoro-styrene, acmc-1crku, 1-fluoro-3-vinyl-benzene, 1-fluoro-3-vinylbenzene #, 3-fluorostyrene, contains 4-tert-butylcatechol as inhibitor
PubChem CID 67691
IUPAC Name 1-ethenyl-3-fluorobenzene
SMILES C=CC1=CC(=CC=C1)F

Specifications

Color Yellow
Boiling Point 41°C
Quantity 5 g
UN Number 1993
Formula Weight 122.14
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Fluorostyrene (stabilized with TBC)
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.