accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 350042-00-1

IUPAC Name: 6-(5-{[2,2'-bipyridin]-6-yl}-1,1-dimethyl-3,4-diphenyl-1H-silol-2-yl)-2,2'-bipyridine
Synonyms: 6-(5-{[2,2'-bipyridin]-6-yl}-1,1-dimethyl-3,4-diphenylsilol-2-yl)-2,2'-bipyridine
Molecular Weight (g/mol): 570.77
Molecular Formula: C38H30N4Si
InChi Key: PEISKVGQULXWNZ-UHFFFAOYSA-N
SMILES: C[Si]1(C)C(=C(C(=C1C1=CC=CC(=N1)C1=CC=CC=N1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC(=N1)C1=CC=CC=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results