accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2NOH2NHNNH2OHSOHOO

CAS RN 35011-47-3

IUPAC Name: 2,5,6-triamino-1,4-dihydropyrimidin-4-one; sulfuric acid
Synonyms: 2,5,6-triamino-1H-pyrimidin-4-one; sulfuric acid
Molecular Weight (g/mol): 239.21
Molecular Formula: C4H9N5O5S
InChi Key: RSKNEEODWFLVFF-UHFFFAOYSA-N
SMILES: OS(O)(=O)=O.NC1=NC(=O)C(N)=C(N)N1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Grade 
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)