accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 351-83-7

IUPAC Name: N-(4-fluorophenyl)acetamide
Synonyms: 4-fluoroacetanilide
Molecular Weight (g/mol): 153.16
Molecular Formula: C8H8FNO
InChi Key: JHEFOJNPLXSWNZ-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(F)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results