accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 351027-75-3

IUPAC Name: N-(2,4,6-trichlorophenyl)benzenesulfonamide
Synonyms: N-(2,4,6-trichlorophenyl)benzenesulfonamide
Molecular Weight (g/mol): 336.61
Molecular Formula: C12H8Cl3NO2S
InChi Key: KVDKXSWOSZISRI-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(NS(=O)(=O)C2=CC=CC=C2)C(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results