accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CONOF

CAS RN 351410-62-3

IUPAC Name: 5-fluoro-2-methoxypyridine-3-carbaldehyde
Synonyms: 5-fluoro-2-methoxypyridine-3-carbaldehyde
Molecular Weight (g/mol): 155.13
Molecular Formula: C7H6FNO2
InChi Key: YAGPZRLCMRMSFP-UHFFFAOYSA-N
SMILES: COC1=NC=C(F)C=C1C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results