accessibility menu, dialog, popup

UserName

Viewing profile as user:
BNHOOH

CAS RN 351422-72-5

IUPAC Name: [3-(pyridin-3-yl)phenyl]boronic acid
Synonyms: 3-(pyridin-3-yl)phenylboronic acid
Molecular Weight (g/mol): 199.02
Molecular Formula: C11H10BNO2
InChi Key: SSVQXHWHJJERAP-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)C1=CN=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results