accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNOHONH2

CAS RN 35146-32-8

IUPAC Name: 2-amino-3-[1-(triphenylmethyl)-1H-imidazol-4-yl]propanoic acid
Synonyms: 2-amino-3-[1-(triphenylmethyl)imidazol-4-yl]propanoic acid
Molecular Weight (g/mol): 397.48
Molecular Formula: C25H23N3O2
InChi Key: BSZQZNOAYQCQFZ-UHFFFAOYNA-N
SMILES: NC(CC1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)