Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Trityl-L-histidine, 98%

Catalog No. AAH6277906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6277906 5 g
AAH6277903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6277906 Supplier Thermo Scientific Chemicals Supplier No. H6277906
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 35146-32-8 | C25H23N3O2 | 397.478 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35146-32-8
Molecular Formula C25H23N3O2
Molecular Weight (g/mol) 397.478
MDL Number MFCD00153444
InChI Key BSZQZNOAYQCQFZ-QHCPKHFHSA-N
Synonym h-his trt-oh, 1-triphenylmethyl-l-histidine, l-histidine, 1-triphenylmethyl, s-2-amino-3-1-trityl-1h-imidazol-4-yl propanoic acid, 2s-2-amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, 1-trityl-l-histidine, pubchem18948, n-im-trityl-l-histidine, n im-trityl-l-histidine
PubChem CID 7408287
IUPAC Name (2S)-2-amino-3-(1-tritylimidazol-4-yl)propanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)N

Specifications

Melting Point 210°C (decomposition)
Beilstein 767244
Quantity 5 g
Formula Weight 397.47
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Trityl-L-histidine
TSCA No
storageNote1 -30° to -10°C
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.