accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 351521-78-3

IUPAC Name: 4-[9-(3,4-dihydroxyphenyl)-9H-fluoren-9-yl]benzene-1,2-diol
Synonyms: 4-[9-(3,4-dihydroxyphenyl)fluoren-9-yl]benzene-1,2-diol
Molecular Weight (g/mol): 382.42
Molecular Formula: C25H18O4
InChi Key: RAIOQXXWEGZKAI-UHFFFAOYSA-N
SMILES: OC1=C(O)C=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(O)=C(O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results