Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

9,9-Bis(3,4-dihydroxyphenyl)fluorene 98.0+%, TCI America™
SDP

Catalog No. B46655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46655G 5 g
B46651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46655G Supplier TCI America Supplier No. B46655G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 351521-78-3
Molecular Formula C25H18O4
Molecular Weight (g/mol) 382.42
MDL Number MFCD31618097
InChI Key RAIOQXXWEGZKAI-UHFFFAOYSA-N
Synonym 4,4′-(9H-Fluorene-9,9-diyl)bis(benzene-1,2-diol)
PubChem CID 24778256
IUPAC Name 4-[9-(3,4-dihydroxyphenyl)-9H-fluoren-9-yl]benzene-1,2-diol
SMILES OC1=C(O)C=C(C=C1)C1(C2=CC=CC=C2C2=CC=CC=C12)C1=CC(O)=C(O)C=C1

Specifications

Melting Point 183°C
Color White
Quantity 5 g
Formula Weight 382.42
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,9-Bis(3,4-dihydroxyphenyl)fluorene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.