accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOCH3CH3(Z)

CAS RN 35154-45-1

IUPAC Name: (3Z)-hex-3-en-1-yl 3-methylbutanoate
Synonyms: cis-3-hexenyl isovalerate
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: AIQLNKITFBJPFO-WAYWQWQTSA-N
SMILES: CC\C=C/CCOC(=O)CC(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results