Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

cis-3-Hexenyl Isovalerate 98.0+%, TCI America™
SDP

Catalog No. I030725ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I030725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. I030725ML Supplier TCI America Supplier No. I030725ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35154-45-1
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.28
MDL Number MFCD00036533
InChI Key AIQLNKITFBJPFO-WAYWQWQTSA-N
Synonym Isovaleric Acid cis-3-Hexenyl Ester
PubChem CID 5367681
IUPAC Name (3Z)-hex-3-en-1-yl 3-methylbutanoate
SMILES CC\C=C/CCOC(=O)CC(C)C

Specifications

Color Colorless
Boiling Point 199°C
Quantity 25 mL
Formula Weight 184.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material cis-3-Hexenyl Isovalerate
EINECSNumber (2)-0760
RTECSNumber NY1505000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.