accessibility menu, dialog, popup

UserName

Viewing profile as user:
MgBr

CAS RN 35166-78-0

IUPAC Name: bromo(cyclohexylmethyl)magnesium
Synonyms: bromo(cyclohexylmethyl)magnesium
Molecular Weight (g/mol): 201.39
Molecular Formula: C7H13BrMg
InChi Key: ZBCIOGFGZRNNEC-UHFFFAOYSA-M
SMILES: Br[Mg]CC1CCCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results