accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3N

CAS RN 35182-51-5

IUPAC Name: 4-(pentan-3-yl)pyridine
Synonyms: pyridine, 4-(1-ethylpropyl)-
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: YHXYCIZBTSECRP-UHFFFAOYSA-N
SMILES: CCC(CC)C1=CC=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results