Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(3-Pentyl)pyridine 95.0+%, TCI America™
SDP

Catalog No. P097025ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P097025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P097025ML Supplier TCI America Supplier No. P097025ML

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 35182-51-5
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00059788
InChI Key YHXYCIZBTSECRP-UHFFFAOYSA-N
Synonym 4-(1-Ethylpropyl)pyridine, 3-(4-Pyridyl)pentane
PubChem CID 101286
IUPAC Name 4-pentan-3-ylpyridine
SMILES CCC(CC)C1=CC=NC=C1

Specifications

Boiling Point 150°C
Quantity 25 mL
Formula Weight 149.24
Percent Purity ≥95.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 4-(3-Pentyl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.