accessibility menu, dialog, popup

UserName

Viewing profile as user:
O

CAS RN 35185-96-7

IUPAC Name: 11-oxatricyclo[6.2.1.0²,⁷]undeca-2,4,6-triene
Synonyms: 11-oxatricyclo[6.2.1.0²,⁷]undeca-2,4,6-triene
Molecular Weight (g/mol): 146.19
Molecular Formula: C10H10O
InChi Key: SPELXJYTVLDKGB-UHFFFAOYNA-N
SMILES: C1CC2OC1C1=CC=CC=C21
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results