Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Epoxy-1,2,3,4-tetrahydronaphthalene 98%, Thermo Scientific™

Catalog No. AC210380250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC210380250 25 g
AC210380050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC210380250 Supplier Thermo Scientific Chemicals Supplier No. 210380250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35185-96-7
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.189
InChI Key SPELXJYTVLDKGB-UHFFFAOYSA-N
Synonym 1,4-epoxy-1,2,3,4-tetrahydronaphthalene, 1,4-epoxynaphthalene, 1,2,3,4-tetrahydro, 1,2,3,4-tetrahydro-1,4-epoxynaphthalene, 11-oxatricyclo 6.2.1.0 2 ,? undeca-2,4,6-triene, 1,4-epoxytetralin, tetrahydro-1,4-epoxynaphthalene, 1,4-epoxynaphthalene,1,2,3,4-tetrahydro, 1,2,3,4-tetrahydronaphthalene 1,4-endoxide, 1,2,3,4-tetrahydronaphthalene-1,4-endo-oxide, 1,4-epoxy-1,2,3,4-tetrahydro-naphthalene
PubChem CID 141969
SMILES C1CC2C3=CC=CC=C3C1O2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.09
Infrared Spectrum Authentic
Refractive Index 1.5550 to 1.5570 (20°C, 589nm)
Quantity 25 g
Beilstein 17, IV, 514
Formula Weight 146.19
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 1,4-Epoxy-1,2,3,4-tetrahydronaphthalene, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.