CAS RN 3519-82-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ NFAT Activation Inhibitor III
The NFAT Activation Inhibitor III, also referenced under CAS 3519-82-2, controls the biological activity of NFAT. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.