Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ NFAT Activation Inhibitor III

Catalog No. 4804035MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-040-35MG 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-040-35MG Supplier MilliporeSigma™ Supplier No. 4804035MG
Add to Cart
Add to Cart

The NFAT Activation Inhibitor III, also referenced under CAS 3519-82-2, controls the biological activity of NFAT. This small molecule/inhibitor is primarily used for Inflammation/Immunology applications.

Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Description 95% by HPLC
Target Proteins Calcineurin-NFAT
CAS 3519-82-2
Molecular Formula C20H12O2
Formula Weight 284.3g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.