accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 35192-74-6

IUPAC Name: sodium nonane-1-sulfonate
Synonyms: sodium nonane-1-sulfonate
Molecular Weight (g/mol): 230.3
Molecular Formula: C9H19NaO3S
InChi Key: RUYRDULZOKULPK-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCS([O-])(=O)=O
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (3)
  • (3)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (3)
  • (3)
  • (4)