accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3OOHNHOO

CAS RN 352351-63-4

IUPAC Name: 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(2-methylphenyl)propanoic acid
Synonyms: 2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(2-methylphenyl)propanoic acid
Molecular Weight (g/mol): 401.46
Molecular Formula: C25H23NO4
InChi Key: GYFMRMRGTNDDAT-UHFFFAOYNA-N
SMILES: CC1=CC=CC=C1CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)