accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3NCH3H3COClNOO

CAS RN 352443-08-4

IUPAC Name: 4-chloro-3-nitro-N,N-bis(propan-2-yl)benzamide
Synonyms: 4-chloro-N,N-diisopropyl-3-nitrobenzamide
Molecular Weight (g/mol): 284.74
Molecular Formula: C13H17ClN2O3
InChi Key: HGKGYXFSDLNRFZ-UHFFFAOYSA-N
SMILES: CC(C)N(C(C)C)C(=O)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results