accessibility menu, dialog, popup

UserName

Viewing profile as user:
CBrFZnI

CAS RN 352530-44-0

IUPAC Name: 4-bromo-2-fluorobenzen-1-ide; iodozincylium
Synonyms: 4-bromo-2-fluorobenzen-1-ide; iodozincylium
Molecular Weight (g/mol): 366.28
Molecular Formula: C6H3BrFIZn
InChi Key: GNZHZNSHJWDVSO-UHFFFAOYSA-M
SMILES: [Zn+]I.FC1=CC(Br)=CC=[C-]1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results