accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 352535-82-1

IUPAC Name: (3-chloro-2-fluorophenyl)boronic acid
Synonyms: 3-chloro-2-fluorophenylboronic acid
Molecular Weight (g/mol): 174.36
Molecular Formula: C6H5BClFO2
InChi Key: SUYRGLRWMPEARP-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(Cl)=C1F
Molecular Weight (g/mol) 
  • (1)
  • (1)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results