CAS RN 35280-78-5
IUPAC Name:
4-({[4-(hexyloxy)phenyl]methylidene}amino)benzonitrile
Synonyms:
4-({[4-(hexyloxy)phenyl]methylidene}amino)benzonitrile
Molecular Weight (g/mol):
306.41
Molecular Formula:
C20H22N2O
InChi Key:
YABQOLANVLHEPV-UHFFFAOYSA-N
SMILES:
CCCCCCOC1=CC=C(C=NC2=CC=C(C=C2)C#N)C=C1
1
–
1
of
1
results
1
4'-Hexyloxybenzylidene-4-cyanoaniline 98.0+%, TCI America™
CAS: 35280-78-5 Molecular Formula: C20H22N2O Molecular Weight (g/mol): 306.41 MDL Number: MFCD00059589 InChI Key: YABQOLANVLHEPV-UHFFFAOYSA-N PubChem CID: 118801 IUPAC Name: 4-({[4-(hexyloxy)phenyl]methylidene}amino)benzonitrile SMILES: CCCCCCOC1=CC=C(C=NC2=CC=C(C=C2)C#N)C=C1