Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Hexyloxybenzylidene-4-cyanoaniline 98.0+%, TCI America™
SDP

Catalog No. H04191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H04191G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H04191G Supplier TCI America Supplier No. H04191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35280-78-5
Molecular Formula C20H22N2O
Molecular Weight (g/mol) 306.41
MDL Number MFCD00059589
InChI Key YABQOLANVLHEPV-UHFFFAOYSA-N
PubChem CID 118801
IUPAC Name 4-({[4-(hexyloxy)phenyl]methylidene}amino)benzonitrile
SMILES CCCCCCOC1=CC=C(C=NC2=CC=C(C=C2)C#N)C=C1

Specifications

Color Yellow
Quantity 1 g
Formula Weight 306.41
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hexyloxybenzylidene-4-cyanoaniline
EINECSNumber (3)-1837
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.