accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaH3COOO

CAS RN 35285-69-9

IUPAC Name: sodium 4-(propoxycarbonyl)benzen-1-olate
Synonyms: sodium propylparaben
Molecular Weight (g/mol): 202.19
Molecular Formula: C10H11NaO3
InChi Key: IXMINYBUNCWGER-UHFFFAOYSA-M
SMILES: [Na+].CCCOC(=O)C1=CC=C([O-])C=C1
Grade 
  • (6)
Percent Purity 
  • (6)
Physical Form 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (6)
  • (5)
  • (6)
  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)